6-Bromo-4-fluoro-1H-indazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A987170-1G A987170-5G
76657-088EA 90.18 USD
76657-088 76657-090
6-Bromo-4-fluoro-1H-indazole-3-carbaldehyde 95%
6-Bromo-4-fluoro-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrFN₂O
MW: 243.0326 g/mol
MDL Number: MFCD07781554
CAS Number: 885520-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR