(S)-tert-Butyl (1-hydroxypent-4-en-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A987165-5G A987165-1G
76657-082EA 41 USD
76657-082 77784-581
(S)-tert-Butyl (1-hydroxypent-4-en-2-yl)carbamate 98%
(S)-tert-Butyl (1-hydroxypent-4-en-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD29042783
CAS Number: 116613-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR