1-Bromo-3-(trimethylsilyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: (3-BRomophenyl)trimethylsilane

A987116-1G A987116-25G A987116-5G
76657-076EA 6 USD
76657-076 76657-078 76657-080
1-Bromo-3-(trimethylsilyl)benzene 95%
1-Bromo-3-(trimethylsilyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₃BrSi
MW: 229.1890 g/mol
MDL Number: MFCD00093882
CAS Number: 17878-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR