5-(4-AMinophenyl)-10,15,20-triphenyl porphine 95%

Supplier: Ambeed
Warning

A987041-100MG A987041-1G A987041-250MG A987041-50MG
76657-068EA 138 USD
76657-068 76657-070 76657-072 76657-074
5-(4-AMinophenyl)-10,15,20-triphenyl porphine 95%
5-(4-AMinophenyl)-10,15,20-triphenyl porphine

Precaución: for Research Use Only

Formula: C₄₄H₃₁N₅
MDL Number: MFCD01074434
CAS Number: 67605-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR