(2S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A986977-1G A986977-5G
76657-060EA 36.08 USD
76657-060 76657-062
(2S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid 98%
(2S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₅NO₄
MW: 391.4596 g/mol
MDL Number: MFCD00191201
CAS Number: 214750-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR