2-(Pyrrolidin-1-yl)pyridin-4-amine 98%

Supplier: Ambeed
Warning

A986964-100MG A986964-250MG
76657-056EA 29 USD
76657-056 76657-058
2-(Pyrrolidin-1-yl)pyridin-4-amine 98%
2-(Pyrrolidin-1-yl)pyridin-4-amine
Formula: C₉H₁₃N₃
MDL Number: MFCD13190680
CAS Number: 35981-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR