N-Benzyl-2-bromo-N-methylacetamide 97%

Supplier: Ambeed
Danger

A986946-100MG A986946-1G A986946-250MG
76657-050EA 22 USD
76657-050 76657-052 76657-054
N-Benzyl-2-bromo-N-methylacetamide 97%
N-Benzyl-2-bromo-N-methylacetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD02751705
CAS Number: 73391-96-5
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR