Ethyl 2-chloro-5-fluoropyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A986893-100MG A986893-250MG
76657-046EA 38 USD
76657-046 76657-048
Ethyl 2-chloro-5-fluoropyrimidine-4-carboxylate 98%
Ethyl 2-chloro-5-fluoropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆ClFN₂O₂
MDL Number: MFCD30742681
CAS Number: 1246632-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR