S-Ethyl-s-phenyl sulfoximine 98%

Supplier: Ambeed
Warning

A986800-100MG A986800-1G A986800-250MG
76657-038EA 43.5 USD
76657-038 76657-040 76657-042
S-Ethyl-s-phenyl sulfoximine 98%
S-Ethyl-s-phenyl sulfoximine

Precaución: for Research Use Only

Formula: C₈H₁₁NOS
MW: 169.2440 g/mol
MDL Number: MFCD09878519
CAS Number: 1889-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR