IKK-16 (hydrochloride) 98%

Supplier: Ambeed
Warning

Synonyms: [4-[[4-(1-benzothiophen-2-yl)pyrimidin-2-yl]amino]phenyl]-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone, hydrochloride, [4-[(4-benzo[b]thien-2-yl-2-pyrimidinyl)amino]phenyl][4-(1-pyrrolidinyl)-1-piperidinyl]-methanone, monohydrochloride, IKK 16 HCl, (4-((4-(Benzo[b]thiophen-2-yl)pyrimidin-2-yl)amino)phenyl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone hydrochloride, IKK Inhibitor VII, IKK 16 hydrochloride

A986748-10MG A986748-1MG A986748-25MG A986748-50MG
76657-026EA 76 USD
76657-026 76657-028 76657-030 76657-032
IKK-16 (hydrochloride) 98%
IKK-16 (hydrochloride)

Precaución: for Research Use Only

Formula: C₂₈H₂₉N₅OS·HCl
MW: 520.1 g/mol
MDL Number: MFCD09971091
CAS Number: 1186195-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR