(3S,5R)-5-(HYdroxymethyl)pyrrolidin-3-ol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A986745-1G
A986745-250MG
A986745-5G
76657-020EA
200.52
USD
76657-020
76657-022
76657-024
(3S,5R)-5-(HYdroxymethyl)pyrrolidin-3-ol hydrochloride 98%
(3S,5R)-5-(HYdroxymethyl)pyrrolidin-3-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂ MW: 153.6073 g/mol |
MDL Number:
MFCD30719389 CAS Number: 1018987-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...