5-Carbomethoxytryptamine hydrochloride 95%

Supplier: Ambeed
Warning

A986677-100MG A986677-250MG
76657-016EA 69 USD
76657-016 76657-018
5-Carbomethoxytryptamine hydrochloride 95%
5-Carbomethoxytryptamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O₂
MW: 254.7127 g/mol
MDL Number: MFCD01321110
CAS Number: 74884-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR