3-(2-Bromoacetamido)phenylboronic acid, pinacol ester 96%

Supplier: Ambeed
Danger

A986448-100MG A986448-1G
76657-006EA 51.99 USD
76657-006 76657-008
3-(2-Bromoacetamido)phenylboronic acid, pinacol ester 96%
3-(2-Bromoacetamido)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BBrNO₃
MW: 340.0206 g/mol
MDL Number: MFCD18783180
CAS Number: 2096331-74-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR