meso-Tetra(4-tert-butylphenyl) porphine 95%

Supplier: Ambeed
Warning

A986304-100MG A986304-1G A986304-250MG A986304-50MG
76656-986EA 127 USD
76656-986 76656-988 76656-990 76656-992
meso-Tetra(4-tert-butylphenyl) porphine 95%
meso-Tetra(4-tert-butylphenyl) porphine

Precaución: for Research Use Only

Formula: C₆₀H₆₂N₄
MW: 839.1611 g/mol
MDL Number: MFCD20502349
CAS Number: 110452-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR