MEso-tetra (p-bromophenyl) porphine 95%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(4-bromophenyl)porphyrin

A986218-1G A986218-250MG A986218-25G A986218-5G
76656-972EA 32 USD
76656-972 76656-974 76656-976 76656-978
MEso-tetra (p-bromophenyl) porphine 95%
MEso-tetra (p-bromophenyl) porphine

Precaución: for Research Use Only

Formula: C₄₄H₂₆Br₄N₄
MW: 930.3201 g/mol
MDL Number: MFCD02114459
CAS Number: 29162-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR