Chloromethyl morpholine-4-carboxylate 97%

Supplier: Ambeed
Danger

A986176-100MG A986176-1G A986176-250MG
76656-966EA 36.08 USD
76656-966 76656-968 76656-970
Chloromethyl morpholine-4-carboxylate 97%
Chloromethyl morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₁₀ClNO₃
MW: 179.6015 g/mol
MDL Number: MFCD20627473
CAS Number: 93765-68-5
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR