Ammonium succinate 98%

Supplier: Ambeed
Warning

A986126-25G A986126-500G A986126-1000G A986126-100G
76656-952EA 7 USD
76656-952 76656-954 77599-364 77784-552
Ammonium succinate 98%
Ammonium succinate

Precaución: for Research Use Only

Formula: C₄H₁₂N₂O₄
MDL Number: MFCD00020534
CAS Number: 2226-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR