1,2-Bis(phenylsulfinyl)ethane 97%

Supplier: Ambeed
Warning

A985942-100MG A985942-1G A985942-250MG A985942-5G
76656-936EA 16.98 USD
76656-936 76656-938 76656-940 77749-824
1,2-Bis(phenylsulfinyl)ethane 97%
1,2-Bis(phenylsulfinyl)ethane

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂S₂
MW: 278.38 g/mol
MDL Number: MFCD11846120
CAS Number: 6099-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR