4-(1-Piperidinyl)pentanoic acid hydrochloride 98%

Supplier: Ambeed
Warning

A985761-1G A985761-250MG
76656-932EA 283 USD
76656-932 76656-934
4-(1-Piperidinyl)pentanoic acid hydrochloride 98%
4-(1-Piperidinyl)pentanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD18483505
CAS Number: 91951-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR