[4-(1-Azepanyl)butyl]amine dihydrochloride 95%

Supplier: Ambeed
Warning

A985374-1G A985374-250MG
76656-926EA 64.72 USD
76656-926 76656-928
[4-(1-Azepanyl)butyl]amine dihydrochloride 95%
[4-(1-Azepanyl)butyl]amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₄Cl₂N₂
MW: 243.22 g/mol
MDL Number: MFCD13186171
CAS Number: 1197880-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR