2-fluoro-3-methyl-5-nitrobenzaldehyde 96%

Supplier: Ambeed
Warning

A985142-100MG A985142-1G A985142-250MG
76656-920EA 50.01 USD
76656-920 76656-922 76656-924
2-fluoro-3-methyl-5-nitrobenzaldehyde 96%
2-fluoro-3-methyl-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FNO₃
MDL Number: MFCD28797464
CAS Number: 1803736-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR