N,N-DImethyl-2-Benzothiazolamine 95%

Supplier: Ambeed
Warning

A985031-100MG A985031-1G A985031-250MG
76656-914EA 13 USD
76656-914 76656-916 76656-918
N,N-DImethyl-2-Benzothiazolamine 95%
N,N-DImethyl-2-Benzothiazolamine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂S
MDL Number: MFCD00613409
CAS Number: 4074-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR