Hexyl laurate 98%

Supplier: Ambeed
Warning

A985016-1G A985016-5G A985016-100MG
76656-910EA 112.46 USD
76656-910 76656-912 77623-280
Hexyl laurate 98%
Hexyl laurate

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₂
MW: 284.4773 g/mol
MDL Number: MFCD09838508
CAS Number: 34316-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR