Methyl 2,2,2-trifluoroethyl carbonate 98%
Supplier: Ambeed
|
Danger
|
A985000-10G
A985000-1G
A985000-25G
A985000-5G
76656-900EA
28
USD
76656-900
76656-902
76656-904
76656-906
Methyl 2,2,2-trifluoroethyl carbonate 98%
Methyl 2,2,2-trifluoroethyl carbonate
Precaución: for Research Use Only
Formula:
C₄H₅F₃O₃ |
MDL Number:
MFCD28054295 CAS Number: 156783-95-8 UN: 3272 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...