2-((tert-Butylamino)methyl)-5-((7-chloroquinolin-4-yl)amino)phenol dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
A984981-10MG
A984981-1MG
A984981-2MG
A984981-50MG
A984981-5MG
76656-890EA
62
USD
76656-890
76656-892
76656-894
76656-896
76656-898
2-((tert-Butylamino)methyl)-5-((7-chloroquinolin-4-yl)amino)phenol dihydrochloride 98%
2-((tert-Butylamino)methyl)-5-((7-chloroquinolin-4-yl)amino)phenol dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₀H₂₄Cl₃N₃O |
MDL Number:
MFCD16627991 CAS Number: 1010411-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...