2-Nitro-4-tert-pentylphenol 95%

Supplier: Ambeed
Warning

A984953-1G A984953-250MG A984953-5G
76656-884EA 18 USD
76656-884 76656-886 76656-888
2-Nitro-4-tert-pentylphenol 95%
2-Nitro-4-tert-pentylphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD16742720
CAS Number: 91247-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR