N,N-DIethyl-6-methyl-5-nitro-2-pyridinamine 95%

Supplier: Ambeed
Warning

A984862-10G A984862-1G A984862-5G
76656-878EA 136.86 USD
76656-878 76656-880 76656-882
N,N-DIethyl-6-methyl-5-nitro-2-pyridinamine 95%
N,N-DIethyl-6-methyl-5-nitro-2-pyridinamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₂
MW: 209.2450 g/mol
MDL Number: MFCD11974904
CAS Number: 28489-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR