2-Bromo-4-chloro-5-nitrophenol 95%

Supplier: Ambeed
Danger

A984795-250MG A984795-1G A984795-5G
76656-876EA 28 USD
76656-876 77599-356 77784-536
2-Bromo-4-chloro-5-nitrophenol 95%
2-Bromo-4-chloro-5-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₃
MW: 252.4499 g/mol
MDL Number: MFCD30534472
CAS Number: 1073437-11-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR