3-Methylpyrrole-2,5-dione 95%

Supplier: Ambeed
Warning

A984754-100MG A984754-250MG A984754-5G
76656-870EA 100.79 USD
76656-870 76656-872 76656-874
3-Methylpyrrole-2,5-dione 95%
3-Methylpyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MW: 111.0987 g/mol
MDL Number: MFCD10699598
CAS Number: 1072-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR