4-Chloro-N-methylbutan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A984665-100MG A984665-250MG
76656-862EA 28 USD
76656-862 76656-864
4-Chloro-N-methylbutan-1-amine hydrochloride 95%
4-Chloro-N-methylbutan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₃Cl₂N
MDL Number: MFCD19232263
CAS Number: 365432-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR