2,2'-(Propane-2,2-diylbis(oxy))diethanamine 98%

Supplier: Ambeed
Warning

A984661-10G A984661-1G A984661-250MG
76656-856EA 413.76 USD
76656-856 76656-858 76656-860
2,2'-(Propane-2,2-diylbis(oxy))diethanamine 98%
2,2'-(Propane-2,2-diylbis(oxy))diethanamine

Precaución: for Research Use Only

Formula: C₇H₁₈N₂O₂
MW: 162.2300 g/mol
MDL Number: MFCD20694614
CAS Number: 127090-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR