2-Bromo-2-chloro-1,1-biphenyl 95%

Supplier: Ambeed
Warning

A984657-1G A984657-250MG A984657-5G A984657-25G
76656-850EA 148.53 USD
76656-850 76656-852 76656-854 77599-354
2-Bromo-2-chloro-1,1-biphenyl 95%
2-Bromo-2-chloro-1,1-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrCl
MW: 267.5489 g/mol
MDL Number: MFCD00466291
CAS Number: 107208-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR