(S)-tert-Butyl 2-Methyl-4-oxopyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A984633-100MG A984633-250MG A984633-1G
76656-846EA 67 USD
76656-846 76656-848 77784-532
(S)-tert-Butyl 2-Methyl-4-oxopyrrolidine-1-carboxylate 95%
(S)-tert-Butyl 2-Methyl-4-oxopyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.2469 g/mol
MDL Number: MFCD23381132
CAS Number: 1374673-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR