1-Boc-piperidine-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid
A984409-100G
A984409-25G
A984409-500G
76656-832EA
21.22
USD
76656-832
76656-834
76656-836
1-Boc-piperidine-4-carboxylic acid 98%
1-Boc-piperidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ MW: 229.3 g/mol |
MDL Number:
MFCD00076999 CAS Number: 84358-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...