3-(2-Methoxyethoxy)prop-1-yne 95%

Supplier: Ambeed
Danger

A984334-5G A984334-1G
76656-824EA 602.6 USD
76656-824 77573-940
3-(2-Methoxyethoxy)prop-1-yne 95%
3-(2-Methoxyethoxy)prop-1-yne

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.1424 g/mol
MDL Number: MFCD13192005
CAS Number: 31995-08-1
UN: 3271
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR