tert-Butyl oxirane-2-carboxylate 95%

Supplier: Ambeed
Danger

A984320-1G A984320-5G
76656-816EA 26.53 USD
76656-816 77595-582
tert-Butyl oxirane-2-carboxylate 95%
tert-Butyl oxirane-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
MDL Number: MFCD24501779
CAS Number: 92223-80-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR