2-(4-Methylbenzyl)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A983908-100G A983908-25G A983908-5G
76656-804EA 55 USD
76656-804 76656-806 76656-808
2-(4-Methylbenzyl)isoindoline-1,3-dione 97%
2-(4-Methylbenzyl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MDL Number: MFCD00023079
CAS Number: 101207-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR