3-Phenoxybenzyl acrylate 97% + stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A983747-100MG A983747-1G A983747-250MG A983747-5G
76656-786EA 15.92 USD
76656-786 76656-788 76656-790 76656-792
3-Phenoxybenzyl acrylate 97% + stabilized
3-Phenoxybenzyl acrylate

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₃
MW: 254.2806 g/mol
CAS Number: 409325-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR