1H-1,2,3-Triazole-4,5-dicarbonitrile 97%

Supplier: Ambeed
Warning

A983745-100MG A983745-1G A983745-250MG A983745-5G
76656-780EA 119 USD
76656-780 76656-782 76656-784 77784-514
1H-1,2,3-Triazole-4,5-dicarbonitrile 97%
1H-1,2,3-Triazole-4,5-dicarbonitrile

Precaución: for Research Use Only

Formula: C₄HN₅
MDL Number: MFCD13193135
CAS Number: 53817-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR