(4-(2-Hydroxy-2-methylpropyl)piperidin-1-yl)(5-methoxy-1H-indol-2-yl)methanone 98%

Supplier: Ambeed
Warning

Synonyms: ASP-9521

A983598-10MG A983598-1MG A983598-50MG A983598-5MG A983598-100MG A983598-1G A983598-250MG
76656-764EA 42 USD
76656-764 76656-766 76656-768 76656-770 77784-511 77784-512 77784-513
(4-(2-Hydroxy-2-methylpropyl)piperidin-1-yl)(5-methoxy-1H-indol-2-yl)methanone 98%
(4-(2-Hydroxy-2-methylpropyl)piperidin-1-yl)(5-methoxy-1H-indol-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂O₃
MDL Number: MFCD30532740
CAS Number: 1126084-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR