Ferric oxalate 97%

Supplier: Ambeed
Warning

A983579-1000G A983579-25G
76656-760EA 89.12 USD
76656-760 76656-762
Ferric oxalate 97%
Ferric oxalate

Precaución: for Research Use Only

Formula: C₆Fe₂O₁₂
MW: 375.7470 g/mol
MDL Number: MFCD00054430
CAS Number: 2944-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR