Methyl 2-(4-bromo-5-methylpyridin-2-yl)acetate 97%

Supplier: Ambeed
Warning

A983529-100MG A983529-1G A983529-250MG
76656-754EA 117.76 USD
76656-754 76656-756 76656-758
Methyl 2-(4-bromo-5-methylpyridin-2-yl)acetate 97%
Methyl 2-(4-bromo-5-methylpyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.0900 g/mol
MDL Number: MFCD28745966
CAS Number: 1805948-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR