4,4-Dimethylcyclopentane-1,3-dione 98%

Supplier: Ambeed
Warning

A983503-100MG A983503-1G A983503-250MG
76656-748EA 77.45 USD
76656-748 76656-750 76656-752
4,4-Dimethylcyclopentane-1,3-dione 98%
4,4-Dimethylcyclopentane-1,3-dione

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.1531 g/mol
MDL Number: MFCD24688183
CAS Number: 4683-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR