4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A983458-100MG
A983458-1G
A983458-250MG
A983458-5G
76656-740EA
71.09
USD
76656-740
76656-742
76656-744
76656-746
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzaldehyde 97%
4,4',4'',4'''-(Ethene-1,1,2,2-tetrayl)tetrabenzaldehyde
Precaución: for Research Use Only
Formula:
C₃₀H₂₀O₄ MW: 444.4774 g/mol |
MDL Number:
MFCD31700764 CAS Number: 2170451-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...