(S)-Methyl 2-((tert-butoxycarbonyl)amino)-2-(4-hydroxyphenyl)acetate 98%

Supplier: Ambeed
Warning

A983446-100MG A983446-1G A983446-250MG A983446-5G
76656-724EA 34 USD
76656-724 76656-726 76656-728 76656-730
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-2-(4-hydroxyphenyl)acetate 98%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-2-(4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₅
MDL Number: MFCD29077614
CAS Number: 225517-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR