Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetate 96%

Supplier: Ambeed
Warning

A983054-100MG A983054-1G
76656-718EA 25 USD
76656-718 76656-720
Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetate 96%
Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₅
MDL Number: MFCD22384993
CAS Number: 141518-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR