3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A983038-100MG A983038-1G A983038-250MG A983038-25G A983038-5G
76656-708EA 21.22 USD
76656-708 76656-710 76656-712 76656-714 76656-716
3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%
3-(4-Hydroxy-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₄
MW: 260.2454 g/mol
MDL Number: MFCD29054753
CAS Number: 1061604-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR