2-Amino-4,5-dimethoxyphenol hydrochloride 96%

Supplier: Ambeed
Warning

A982827-100MG A982827-250MG
76656-686EA 85 USD
76656-686 76656-688
2-Amino-4,5-dimethoxyphenol hydrochloride 96%
2-Amino-4,5-dimethoxyphenol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO₃
MDL Number: MFCD31923000
CAS Number: 2368871-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR