2,2-Dimethylbenzo[d][1,3]dioxole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A982659-100MG A982659-1G A982659-250MG
76656-684EA 82.99 USD
76656-684 77573-932 77784-495
2,2-Dimethylbenzo[d][1,3]dioxole-5-carbaldehyde 97%
2,2-Dimethylbenzo[d][1,3]dioxole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD20488897
CAS Number: 63124-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR