HS-1371 97%

Supplier: Ambeed
Warning

Synonyms: EX-A2073, 4-(4-methylphenoxy)-7-(1-piperidin-4-ylpyrazol-4-yl)quinoline, 7-(1-(Piperidin-4-yl)-1H-pyrazol-4-yl)-4-(p-tolyloxy)quinoline, HS1371

A982329-10MG A982329-1MG A982329-25MG A982329-50MG A982329-5MG
76656-674EA 58.35 USD
76656-674 76656-676 76656-678 76656-680 76656-682
HS-1371 97%
HS-1371

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₂₄O
MW: 384.47 g/mol
MDL Number: MFCD31813738
CAS Number: 2158197-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR